Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitroethanol 98%

2-Nitroethanol 98%

Bottle
Catalog # APOSOR924474-25G
Irritant
Supplier:   Apollo Scientific
2-Nitroethanol 98%
Catalog # APOSOR924474-25G
Supplier:  Apollo Scientific
CAS Number:  625-48-9
2-Nitroethanol 98%

Bottle

CAS Number:  625-48-9
Price
98,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:625-48-9
  • Formula:C₂H₅NO₃
  • MDL Number:MFCD00007405
  • Molecular Weight:91,065 g/mol
  • EINECS:210-895-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications