0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2-Mercaptopyrimidine
BP0000733991
2-Mercaptopyrimidine
Bottle
APOSOR1010512-100G
:
Apollo Scientific
2-Mercaptopyrimidine
APOSOR1010512-100G
:
Apollo Scientific
:
2-Mercaptopyrimidine
Bottle
:
181,00 €
1 * 100 g
-
+
Pack type:
Bottle