Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Diaminononane

1,9-Diaminononane

Bottle
Catalog # APOSOR10105-100G
Corrosive
New Product
Supplier:   Apollo Scientific
1,9-Diaminononane
Catalog # APOSOR10105-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  646-24-2
1,9-Diaminononane

Bottle

CAS Number:  646-24-2
Price
1 710,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,9-Nonanediamine, Nonamethylendiamine
  • CAS Number:646-24-2
  • Formula:C₉H₂₂N₂
  • MDL Number:MFCD00008251
  • Molecular Weight:158.29 g/mol
  • Boiling Pt:258…259 °C
  • Melting Pt:35…40 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:211-470-3
  • UN:3259
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications