Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hexylthiophene 98%

2-Hexylthiophene 98%

Bottle
Catalog # APOSOR1010458-25G
Supplier:   Apollo Scientific
2-Hexylthiophene 98%
Catalog # APOSOR1010458-25G
Supplier:  Apollo Scientific
CAS Number:  18794-77-9
2-Hexylthiophene 98%

Bottle

CAS Number:  18794-77-9
Price
59,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:18794-77-9
  • Formula:C₁₀H₁₆S
  • MDL Number:MFCD00022535
  • Molecular Weight:168,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications