Order Entry
Export
Orders LinkContactUsLinkComponent
Vinylferrocene 98%

Vinylferrocene 98%

Bottle
Catalog # APOSIN10648-25G
Flammable
Supplier:   Apollo Scientific
Vinylferrocene 98%
Catalog # APOSIN10648-25G
Supplier:  Apollo Scientific
CAS Number:  1271-51-8
Vinylferrocene 98%

Bottle

CAS Number:  1271-51-8
Price
343,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Inorganic salt or catalyst.

  • Store at 2 to 8 °C
  • CAS Number:1271-51-8
  • Formula:C₁₂H₁₂Fe
  • MDL Number:MFCD00001434
  • Molecular Weight:212,07 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:215-041-1
  • UN:1325
Specifications

Specifications

  • Pack type:
    Bottle

Specifications