Vinylferrocene 98%
Bottle
Catalog # APOSIN10648-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Inorganic salt or catalyst.
- Store at 2 to 8 °C
- CAS Number:1271-51-8
- Formula:C₁₂H₁₂Fe
- MDL Number:MFCD00001434
- Molecular Weight:212,07 g/mol
- Storage Temperature:Refrigerator
- EINECS:215-041-1
- UN:1325
Specifications
Specifications
- Pack type:Bottle



