Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Diaminopyridine

2,3-Diaminopyridine

Bottle
Catalog # APOSOR0033-100G
Toxic
New Product
Supplier:   Apollo Scientific
2,3-Diaminopyridine
Catalog # APOSOR0033-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  452-58-4
2,3-Diaminopyridine

Bottle

CAS Number:  452-58-4
Price
228,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Pyridine-2,3-diyldiamine
  • CAS Number:452-58-4
  • Formula:C₅H₇N₃
  • MDL Number:MFCD00006319
  • Molecular Weight:109.13 g/mol
  • Boiling Pt:195 °C/55mmHg
  • Melting Pt:110…115 °C
  • Storage Temperature:Ambient
  • EINECS:207-200-9
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications