Order Entry
Export
Orders LinkContactUsLinkComponent
DL-alpha-Tocopherol 97%

DL-alpha-Tocopherol 97%

Bottle
Catalog # APOSBIT3091-100G
Irritant
Supplier:   Apollo Scientific
DL-alpha-Tocopherol 97%
Catalog # APOSBIT3091-100G
Supplier:  Apollo Scientific
CAS Number:  10191-41-0
DL-alpha-Tocopherol 97%

Bottle

CAS Number:  10191-41-0
Price
94,10 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at −20 °C
Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • MDL Number:MFCD00072051
  • Molecular Weight:430,70609 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle

Specifications