Order Entry
Export
Orders LinkContactUsLinkComponent
 

L(-)-Tryptophan ≥99.5%

Bottle
Catalog # APOSBIT0720-250G
Irritant
New Product
Supplier:   Apollo Scientific
L(-)-Tryptophan ≥99.5%
Catalog # APOSBIT0720-250G
Supplier:  Apollo Scientific
New Product
CAS Number:  73-22-3
L(-)-Tryptophan ≥99.5%

Bottle

CAS Number:  73-22-3
Price
33,30 €
Log in for availability
1 * 250 g
Product Details & Documents

USP, pyrogen free

Synonyms:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS Number:73-22-3
  • Formula:C₁₁H₁₂N₂O₂
  • MDL Number:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Melting Pt:280…285 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-795-6

 

Specifications

Specifications

  • Pack type:
    Bottle

Specifications