L(-)-Tryptophan ≥99.5%
Bottle
Catalog # APOSBIT0720-2.5KG
New Product
Supplier: Apollo Scientific
L(-)-Tryptophan ≥99.5%
Catalog # APOSBIT0720-2.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 73-22-3
Jump to:
Product Details & Documents
USP, pyrogen free
Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
- CAS Number:73-22-3
- Formula:C₁₁H₁₂N₂O₂
- MDL Number:MFCD00064340
- Molecular Weight:204.23 g/mol
- Melting Pt:280…285 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-795-6
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle



