Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Proline ≥99%

L(-)-Proline ≥99%

Bottle
Catalog # APOSBIP0717-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
L(-)-Proline ≥99%
Catalog # APOSBIP0717-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  147-85-3
L(-)-Proline ≥99%

Bottle

CAS Number:  147-85-3
Price
163,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

USP, pyrogen free

Synonyms:  (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
  • CAS Number:147-85-3
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00064318
  • Molecular Weight:115.13 g/mol
  • Melting Pt:228 °C (dec.)
  • Density:1.36 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:205-702-2

Documents

Specifications

Specifications

  • Pack type:
    Bottle

Specifications