L(-)-Proline ≥99%
Bottle
Catalog # APOSBIP0717-10KG
New Product
Supplier: Apollo Scientific
L(-)-Proline ≥99%
Catalog # APOSBIP0717-10KG
Supplier: Apollo Scientific
New Product
CAS Number: 147-85-3
Jump to:
Product Details & Documents
USP, pyrogen free
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Number:147-85-3
- Formula:C₅H₉NO₂
- MDL Number:MFCD00064318
- Molecular Weight:115.13 g/mol
- Melting Pt:228 °C (dec.)
- Density:1.36 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:205-702-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle



