0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Orange G
BP0000728044
Orange G
APOSBIGE1010-5G
New Product
:
Apollo Scientific
Orange G
APOSBIGE1010-5G
:
Apollo Scientific
New Product
:
Orange G
:
21,80 €
1 * 5 g
-
+