Orders LinkContactUsLinkComponent
17-AAG
Bottle
  APOSBIA4307-5MG
New Product
17-AAG
  APOSBIA4307-5MG
New Product
 :  75747-14-7
17-AAG
Bottle
 :  75747-14-7
53,60 €
1 * 5 mg
Hsp90 inhibitor.
 :  17-(Allylamino)-17-demethoxygeldanamycin, Tanespimycin, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
  •  :75747-14-7
  •  :C₃₁H₄₃N₃O₈
  •  :MFCD04973892
  •  :585.7 g/mol
  •  :Freezer
  • Pack type:
    Bottle