D-Alanine
Bottle
Catalog # APOSBIA0863-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Chiral intermediate for pharmaceuticals and agrochemicals.
- Materials for peptides
- USP
- Pyrogen free
Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
- CAS Number:338-69-2
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008077
- Molecular Weight:89.09 g/mol
- Melting Pt:291…295 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-418-1
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle



