Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-Alanine

Bottle
Catalog # APOSBIA0863-10KG
Irritant
Supplier:   Apollo Scientific
D-Alanine
Catalog # APOSBIA0863-10KG
Supplier:  Apollo Scientific
CAS Number:  338-69-2
D-Alanine

Bottle

CAS Number:  338-69-2
Price
854,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

Chiral intermediate for pharmaceuticals and agrochemicals.

  • Materials for peptides
  • USP
  • Pyrogen free
Synonyms:  (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
  • CAS Number:338-69-2
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008077
  • Molecular Weight:89.09 g/mol
  • Melting Pt:291…295 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-418-1

 

Specifications

Specifications

  • Pack type:
    Bottle

Specifications