Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl Gallate 98%

Ethyl Gallate 98%

Bottle
Catalog # APOSOR1013235-100G
Supplier:   Apollo Scientific
Ethyl Gallate 98%
Catalog # APOSOR1013235-100G
Supplier:  Apollo Scientific
CAS Number:  831-61-8
Ethyl Gallate 98%

Bottle

CAS Number:  831-61-8
Price
66,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
  • CAS Number:831-61-8
  • Formula:C₉H₁₀O₅
  • MDL Number:MFCD00016430
  • Molecular Weight:198,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications