Order Entry
Export
Orders LinkContactUsLinkComponent
Benzoic acid

Benzoic acid

Bottle
Catalog # APOSOR9594-500G
Corrosive
Irritant
Health
New Product
Supplier:   Apollo Scientific
Benzoic acid
Catalog # APOSOR9594-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  65-85-0
Benzoic acid

Bottle

CAS Number:  65-85-0
Price
36,20 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:65-85-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00002398
  • Molecular Weight:122.12 g/mol
  • Boiling Pt:249 °C
  • Melting Pt:121…125 °C
  • Density:1.32 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:200-618-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications