0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
5-Hydroxy-2-nitropyridine
5-Hydroxy-2-nitropyridine 95%
5-Hydroxy-2-nitropyridine 95%
5-Hydroxy-2-nitropyridine 95%
Bottle
APOSOR959407-25G
New Product
:
Apollo Scientific
5-Hydroxy-2-nitropyridine 95%
APOSOR959407-25G
:
Apollo Scientific
New Product
:
15206-26-5
Expand image
5-Hydroxy-2-nitropyridine 95%
Bottle
Add to cart
:
15206-26-5
2 190,00 €
Unit of Measure
1 * 25 g
-
+
Add to cart
:
6-nitropyridin-3-ol
:
15206-26-5
:
C5H4N2O3
:
MFCD03032441
:
140.0969 g/mol
:
Ambient
Pack type:
Bottle