Order Entry
Export
Orders LinkContactUsLinkComponent
Nonadecan-10-one 98%

Nonadecan-10-one 98%

Bottle
Catalog # APOSOR95811-5G
Supplier:   Apollo Scientific
Nonadecan-10-one 98%
Catalog # APOSOR95811-5G
Supplier:  Apollo Scientific
CAS Number:  504-57-4
Nonadecan-10-one 98%

Bottle

CAS Number:  504-57-4
Price
72,40 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Caprinone, Dinonyl ketone
  • CAS Number:504-57-4
  • Formula:C₁₉H₃₈O
  • MDL Number:MFCD00009584
  • Molecular Weight:282,5 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications