Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
10-Nonadecanone
BP0000757401
Print
Nonadecan-10-one
Bottle
Catalog # APOSOR95811-100G
Supplier:
Apollo Scientific
Nonadecan-10-one
Catalog # APOSOR95811-100G
Supplier:
Apollo Scientific
CAS Number:
Return to Top
Nonadecan-10-one
Bottle
Specifications
Jump to:
Specifications
CAS Number:
Price
1 040,00 €
Log in
for availability
1 * 100 g
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications