Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Difluorophenylhydrazine

2,6-Difluorophenylhydrazine

Bottle
Catalog # APOSPC1502-25G
Supplier:   Apollo Scientific
2,6-Difluorophenylhydrazine
Catalog # APOSPC1502-25G
Supplier:  Apollo Scientific
CAS Number:  119452-66-3
2,6-Difluorophenylhydrazine

Bottle

CAS Number:  119452-66-3
Price
712,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (2,6-Difluorophenyl)hydrazine
  • CAS Number:119452-66-3
  • Formula:C₆H₆F₂N₂
  • MDL Number:MFCD06658266
  • Molecular Weight:144.122 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications