Order Entry
Export
Orders LinkContactUsLinkComponent
2,2'-Dipyrrolylmethane 95%

2,2'-Dipyrrolylmethane 95%

Bottle
Catalog # APOSOR932562-25G
Irritant
New Product
Supplier:   Apollo Scientific
2,2'-Dipyrrolylmethane 95%
Catalog # APOSOR932562-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  21211-65-4
2,2'-Dipyrrolylmethane 95%

Bottle

CAS Number:  21211-65-4
Price
423,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Di(1H-pyrrol-2-yl)methane
  • CAS Number:21211-65-4
  • Formula:C₉H₁₀N₂
  • MDL Number:MFCD03931560
  • Molecular Weight:146.19 g/mol
  • Boiling Pt:120 °C/2.25mmHg
  • Melting Pt:74 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications