Order Entry
Export
Orders LinkContactUsLinkComponent
Ac-D-Pro-Oh 98%

Ac-D-Pro-Oh 98%

Bottle
Catalog # APOSOR1012186-500G
Supplier:   Apollo Scientific
Ac-D-Pro-Oh 98%
Catalog # APOSOR1012186-500G
Supplier:  Apollo Scientific
CAS Number:  59785-68-1
Ac-D-Pro-Oh 98%

Bottle

CAS Number:  59785-68-1
Price
429,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  N-Acetyl-D-proline
  • CAS Number:59785-68-1
  • Formula:C₇H₁₁NO₃
  • MDL Number:MFCD20640695
  • Molecular Weight:157,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications