Ac-D-Pro-Oh 98%
Bottle
Catalog # APOSOR1012186-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: N-Acetyl-D-proline
- CAS Number:59785-68-1
- Formula:C₇H₁₁NO₃
- MDL Number:MFCD20640695
- Molecular Weight:157,17 g/mol
Specifications
Specifications
- Pack type:Bottle



