AC-Beta-ALA-OH 97%
Bottle
Catalog # APOSOR1012185-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
- CAS Number:3025-95-4
- Formula:C₅H₉NO₃
- MDL Number:MFCD00037296
- Molecular Weight:131,13 g/mol
Specifications
Specifications
- Pack type:Bottle



