N-Allylurea 98%
Bottle
Catalog # APOSOR80688-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: N-Allylurea
- CAS Number:557-11-9
- Formula:C₄H₈N₂O
- MDL Number:MFCD00007954
- Molecular Weight:100,12 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR80688-500G



