Order Entry
Export
Orders LinkContactUsLinkComponent
N-Allylurea 98%

N-Allylurea 98%

Bottle
Catalog # APOSOR80688-100G
Supplier:   Apollo Scientific
N-Allylurea 98%
Catalog # APOSOR80688-100G
Supplier:  Apollo Scientific
CAS Number:  557-11-9
N-Allylurea 98%

Bottle

CAS Number:  557-11-9
Price
128,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  N-Allylurea
  • CAS Number:557-11-9
  • Formula:C₄H₈N₂O
  • MDL Number:MFCD00007954
  • Molecular Weight:100,12 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications