Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Dichlorononane 98%

1,9-Dichlorononane 98%

Bottle
Catalog # APOSOR80225-500G
New Product
Supplier:   Apollo Scientific
1,9-Dichlorononane 98%
Catalog # APOSOR80225-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  821-99-8
1,9-Dichlorononane 98%

Bottle

CAS Number:  821-99-8
Price
948,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:821-99-8
  • Formula:C₉H₁₈Cl₂
  • MDL Number:MFCD00001024
  • Molecular Weight:197,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications