Order Entry
Export
Orders LinkContactUsLinkComponent
Pyrazolo[1,5-a]pyridine-7-methanamine 1g pack 1 * 1 g

Pyrazolo[1,5-a]pyridine-7-methanamine 1g pack 1 * 1 g

Catalog # APOSOR87447-1G
New Product
Supplier:   Apollo Scientific
Pyrazolo[1,5-a]pyridine-7-methanamine 1g pack 1 * 1 g
Catalog # APOSOR87447-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Pyrazolo[1,5-a]pyridine-7-methanamine 1g pack 1 * 1 g
CAS Number:  
Price
898,00 €
Log in for availability
1 * 1 g
Product Details & Documents