Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC COLUMN HI-5 0.32MM 0.18UM 30M 1 * 1 items
Catalog # HIGCHI02-32-018-30
Supplier:
Avantor
New Product
Expand image
GC COLUMN HI-5 0.32MM 0.18UM 30M 1 * 1 items
Catalog # HIGCHI02-32-018-30
Supplier:
Avantor
New Product