3 components: 20g/l each of Fe Pb Zn in HNO3 5% ,, CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM081332L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product