Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
900MG MGS04, 300MG PSA, 300MG C18, 100MG SILICA, 45MG GCB, PREPACKED IN 15ML TUBES, QUECHERS, 50 PACK 1 * 50 items
HYPES1-15-CN-KIT
:
Thermo Fisher Scientific
New Product
900MG MGS04, 300MG PSA, 300MG C18, 100MG SILICA, 45MG GCB, PREPACKED IN 15ML TUBES, QUECHERS, 50 PACK 1 * 50 items
HYPES1-15-CN-KIT
:
Thermo Fisher Scientific
New Product