Orders LinkContactUsLinkComponent
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Glass test tube 16×100 mm, high borosilicate, PP cap with silicone pad (100 tubes per box)
Catalog # GRANDENTUBE16
Supplier:   GRANT INSTRUMENTS
Accessories for Densitometer for Cell Suspensions
Catalog # GRANDENTUBE16
Supplier:  GRANT INSTRUMENTS
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Glass test tube 16×100 mm, high borosilicate, PP cap with silicone pad (100 tubes per box)
Price
75,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Specifications
  • Description:
    Glass test tube 16×100 mm, high borosilicate, PP cap with silicone pad (100 tubes per box)