Orders LinkContactUsLinkComponent
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Glass Test Tube 18×100 mm. High borosilicate, PP Cap with silicone pad
Catalog # GRANDENTUBE18
Supplier:   GRANT INSTRUMENTS
Accessories for Densitometer for Cell Suspensions
Catalog # GRANDENTUBE18
Supplier:  GRANT INSTRUMENTS
Accessories for Densitometer for Cell Suspensions
Accessories for Densitometer for Cell Suspensions
Glass Test Tube 18×100 mm. High borosilicate, PP Cap with silicone pad
Price
75,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Specifications
  • Description:
    Glass Test Tube 18×100 mm. High borosilicate, PP Cap with silicone pad