Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC COLUMN HI-WAX 0.32MM 0.20UM 50M 1 * 1 items
Catalog # HIGCHI09-32-020-50
Supplier:
Avantor
New Product
Expand image
GC COLUMN HI-WAX 0.32MM 0.20UM 50M 1 * 1 items
Catalog # HIGCHI09-32-020-50
Supplier:
Avantor
New Product