ContactUsLinkComponent
GC COLUMN HI-1 0.32MM 0.17UM 50M 1 * 1 items
Catalog # HIGCHI01-32-017-50
Supplier:  Avantor
New Product
GC COLUMN HI-1 0.32MM 0.17UM 50M 1 * 1 items
Catalog # HIGCHI01-32-017-50
Supplier:  Avantor
New Product