ContactUsLinkComponent
RD Mass Spec Liquid Waste Kit - 3x Canister 10 L, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 200 x 258 x 300 mm - 1x Collecting tray, PP electrostatic conductive, dimensions (W x H x D): 370 x 265 x 570 mm (interio 1 * 1 KIT
Catalog # SCTV116972
Supplier:  S.C.A.T.
New Product
RD Mass Spec Liquid Waste Kit - 3x Canister 10 L, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 200 x 258 x 300 mm - 1x Collecting tray, PP electrostatic conductive, dimensions (W x H x D): 370 x 265 x 570 mm (interio 1 * 1 KIT
Catalog # SCTV116972
Supplier:  S.C.A.T.
New Product