Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Masterflex Transfer Tubing, Flexible PVC, non-DEHP, 1/4 x 1/2, 50 ft 1 * 50 Feet
Catalog # MFLX96505-18
Supplier:
Avantor Fluid Handling
New Product
Expand image
Masterflex Transfer Tubing, Flexible PVC, non-DEHP, 1/4 x 1/2, 50 ft 1 * 50 Feet
Catalog # MFLX96505-18
Supplier:
Avantor Fluid Handling
New Product