ContactUsLinkComponent
Product developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplan tation genetic testing 1 * 1 KIT
Catalog # BIOO4340-0248
Supplier:  BIOO SCIENTIFIC
New Product
Product developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplan tation genetic testing 1 * 1 KIT
Catalog # BIOO4340-0248
Supplier:  BIOO SCIENTIFIC
New Product