NEXTFLEX™ Rapid DNA-seq 2.0 Kits
Catalog # BIOONOVA-5188-01
Supplier: BIOO SCIENTIFIC
New Product
CAS Number:
Specifications
- Description:NEXTFLEX™ Rapid DNA-seq kit 2.0
- Seq:1
- No. of reactions:8
Specifications
About this item
The NEXTFLEX® Rapid DNA-seq 2.0 kit is designed to prepare multiplexed libraries for single or paired-end sequencing using Illumina® platforms from fragmented DNA.
- Complete library prep solution, including size selection beads Up to 1536 UDI barcodes are available separately Convenient and cost-effective package in an 8, 48, or 96 reaction format Simplified workflow Robust genome coverage Reliable performance with GC-bias and sequencing bias mitigation Low input requirement for genomic DNA down to 1 ng Functionally tested with Illumina® sequencing platform Automation compatible with liquid handlers, including Revvity's Sciclone® G3 NGSx workstations, Zephyr® G3 NGS workstation, and BioQule NGS system
The NEXTFLEX® Rapid DNA-seq kit 2.0 produces library preps with a larger number of unique sequencing reads, allowing for robust genome coverage in challenging GC-rich sequences that can introduce bias.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.