Order Entry
Export
Orders LinkContactUsLinkComponent
NEXTFLEX™ Cell Free DNA-Sequence Kits

NEXTFLEX™ Cell Free DNA-Sequence Kits

NEXTFLEX™ cell free DNA-sequencing, 48
Catalog # BIOONOVA-5150-02
Supplier:   Revvity
NEXTFLEX™ Cell Free DNA-Sequence Kits
Catalog # BIOONOVA-5150-02
Supplier:  Revvity
CAS Number:  
NEXTFLEX™ Cell Free DNA-Sequence Kits
NEXTFLEX™ Cell Free DNA-Sequence Kits

NEXTFLEX™ cell free DNA-sequencing, 48

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

The NEXTFLEX™ cell free DNA-sequence kit 2.0 can produce libraries from 10 ng of cell free DNA for circulating tumor DNA (ctDNA) or cell free fetal DNA (cffDNA) analysis, in two hours.

  • Designed for low sample input - 10 ng of input cfDNA required Accelerated workflow with minimised hands-on time Flexible adapter barcode options - up to 1536 unique dual index barcodes are available Compatible with Illumina® sequencing platforms Protocols available for automation on the Sciclone® G3 NGSx, NGSx iQ™ and Zephyr® G3 NGS workstations

The NEXTFLEX™ cell free DNA-sequence kit 2.0 delivers high coverage and reduced bias for Illumina® sequencing applications. The NEXTFLEX™ cell free DNA-sequence kit 2.0 can be used to prepare single, paired-end and multiplexed DNA libraries for sequencing using Illumina® platforms. The availability of up to 1536 unique dual index barcodes facilitates high-throughput applications without compromising data integrity.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ cell free DNA-sequencing
  • No. of reactions:
    48

Specifications