NEXTFLEX™ Cell Free DNA-Sequence Kits
Catalog # BIOONOVA-5150-01
Supplier: BIOO SCIENTIFIC
New Product
CAS Number:
Specifications
- Description:NEXTFLEX™ cell free DNA-sequencing
- Seq:1
- No. of reactions:8
Specifications
About this item
The NEXTFLEX™ cell free DNA-sequence kit 2.0 can produce libraries from 10 ng of cell free DNA for circulating tumor DNA (ctDNA) or cell free fetal DNA (cffDNA) analysis, in two hours.
- Designed for low sample input - 10 ng of input cfDNA required Accelerated workflow with minimised hands-on time Flexible adapter barcode options - up to 1536 unique dual index barcodes are available Compatible with Illumina® sequencing platforms Protocols available for automation on the Sciclone® G3 NGSx, NGSx iQ™ and Zephyr® G3 NGS workstations
The NEXTFLEX™ cell free DNA-sequence kit 2.0 delivers high coverage and reduced bias for Illumina® sequencing applications. The NEXTFLEX™ cell free DNA-sequence kit 2.0 can be used to prepare single, paired-end and multiplexed DNA libraries for sequencing using Illumina® platforms. The availability of up to 1536 unique dual index barcodes facilitates high-throughput applications without compromising data integrity.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.