Order Entry
Export
Orders LinkContactUsLinkComponent
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits

NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits

NEXTFLEX™ Rapid XP V2 DNA-sequence kit, 8
Catalog # BIOONOVA-5149-21
Supplier:   Revvity
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
Catalog # BIOONOVA-5149-21
Supplier:  Revvity
CAS Number:  
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits

NEXTFLEX™ Rapid XP V2 DNA-sequence kit, 8

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

The NEXTFLEX® Rapid XP V2 DNA-sequence kit offers a simplified DNA-seq library prep workflow for Illumina® platforms. Libraries produced using this kit are normalised during library construction.

  • Fully automated normalization and fragmentation Enzymatic fragmentation with end-repair and A-tailing in one reaction Highly efficient ligation with low adapter dimer formation Efficient: Automated on the Sciclone® and Zephyr® NGS workstations for mid-high throughput automation and now on the BioQule™ NGS System for low throughput labs

The NEXTFLEX® Rapid XP V2 DNA-seq kit includes proprietary NEXTFLEX® normalization beads, which provide a consistent mass and sequencing cluster density for all samples in a library pool. This shortens the time needed for quantification and pooling preparation for sequencing in a high throughput lab by up to 3 hours per 96 samples. The NEXTFLEX® Rapid XP V2 DNA-seq kit offers a streamlined workflow with highly efficient ligation, generating low amount of adapter dimers even at low input concentrations.

The NEXTFLEX Rapid XP V2 DNA-Seq kit is compatible with multiple liquid handlers, including the Sciclone G3 NGSx workstation, Sciclone G3 NGSx iQ™ workstation, and Zephyr NGS workstation. For low throughput automation, the NEXTFLEX Rapid XP V2 DNA-Seq kit is also now automated on the BioQule™ NGS System which simplifies both library prep and quantitation. This low-cost, benchtop instrument delivers libraries ready to load into your sequencer with only 15 minutes of hands-on time.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
  • No. of reactions:
    8

Specifications