Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Depletion kit (human / mouse / rat) that is a effective method to remove rRNA contamination from mammalian total RNA 1 * 1 KIT
Catalog # BIOONOVA-512965
Supplier:
BIOO SCIENTIFIC
New Product
Expand image
Depletion kit (human / mouse / rat) that is a effective method to remove rRNA contamination from mammalian total RNA 1 * 1 KIT
Catalog # BIOONOVA-512965
Supplier:
BIOO SCIENTIFIC
New Product