ContactUsLinkComponent
Microscope cover glasses, DIN ISO 8255 compliant, 28 mm Ø, borosilicate glass D 263® M, thickness No. 1.5H (0.17±0.005 mm), CE marked, 100 pcs./cardboard box, 1000 pcs./carton, MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI0117680
Supplier:  MARIENFELD
New Product
Microscope cover glasses, DIN ISO 8255 compliant, 28 mm Ø, borosilicate glass D 263® M, thickness No. 1.5H (0.17±0.005 mm), CE marked, 100 pcs./cardboard box, 1000 pcs./carton, MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI0117680
Supplier:  MARIENFELD
New Product