Order Entry
Export
Orders LinkContactUsLinkComponent
Buffered NaCl Peptone Solution pH 7,0

Buffered NaCl Peptone Solution pH 7,0

Buffered NaCl peptone solution, 7,0, 300 ml, 6
Catalog # LIOF403410
Supplier:   LIOFILCHEM
Buffered NaCl Peptone Solution pH 7,0
Catalog # LIOF403410
Supplier:  LIOFILCHEM
CAS Number:  
Buffered NaCl Peptone Solution pH 7,0

Buffered NaCl peptone solution, 7,0, 300 ml, 6

CAS Number:  
Price
56,00 €
Log in for availability
1 * 6 items
Product Details & Documents

Buffered NaCl Peptone Solution pH 7,0 is used for dissolving, suspending and diluting test samples. Its composition conforms to recommendations in the Harmonised Pharmacopoeias for the microbiological examination of non-sterile products.

Certifications: This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics.

Ordering information: For professional use only.

Specifications

Specifications

  • Pk:
    6
  • Description:
    Buffered NaCl peptone solution
  • Filling:
    300 ml
  • pH value:
    7,0
  • Cat. No.:
    LIOF403410
  • Storage temperature:
    10...25 °C

Specifications