Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR® Flask Brushes

Brush for 50 ml flasks, Black, 44 - 54 mm, Wire
  VWRU17145-052
VWR® Flask Brushes
  VWRU17145-052
VWR® Flask Brushes

Brush for 50 ml flasks, Black, 44 - 54 mm, Wire

 
153,00 €
   
1 * 1 items
 

These brushes with special construction puts the cleaning action directly where it is needed.

The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.

 
  • Bristle colour:
    Black
  • Brush head Ø:
    44 - 54 mm
  • Handle material:
    Wire
  • Description:
    Brush for 50 ml flasks
  • Seq:
    02