Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Flask Brushes

VWR® Flask Brushes

Brush for 25 ml flasks, Black, 44 mm, Wire
Catalog # VWRU17145-050
Supplier:   VWR Collection
VWR® Flask Brushes
Catalog # VWRU17145-050
Supplier:  VWR Collection
VWR® Flask Brushes

Brush for 25 ml flasks, Black, 44 mm, Wire

Price
12,30 €
Log in for availability
1 * 1 items
Product Details & Documents

These brushes with special construction puts the cleaning action directly where it is needed.

The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.

Specifications
  • Bristle colour:
    Black
  • Brush head Ø:
    44 mm
  • Handle material:
    Wire
  • Description:
    Brush for 25 ml flasks
  • Seq:
    01