Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, extended length, 20 µl, -, -, 49,45 mm, Clear, 10 racks, 96 each
Catalog # 613-7414
Supplier:   Corning
Axygen® LTS Compatible Pipette Tips
Catalog # 613-7414
Supplier:  Corning
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, extended length, 20 µl, -, -, 49,45 mm, Clear, 10 racks, 96 each
Price
47,20 €
Log in for availability
1 * 960 items
Product Details & Documents
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
Certifications: ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    20 µl
  • Length:
    49,45 mm
  • Seq:
    10
  • Tip style:
    LTS compatible, extended length
  • Tip colour:
    Clear
  • Filter:
    -